Product Details
- SNP ID
-
rs13482424
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:9192536 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACAGCTATACAGTAGCTTTGGTT[A/G]TAAAAATTGATCACATGAGAAACCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Lmbrd2
PubMed Links
Gene Details
- Gene
- Lmbrd2
- Gene Name
- LMBR1 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177178.3 |
|
Intron |
|
|
NP_796152.2 |
View Full Product Details