Product Details
- SNP ID
-
rs13475709
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:6655964 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTCATTCTATTCTCTGGTCTCCA[C/G]GTACTACTCAGCACATTGCAAATGT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
St18
PubMed Links
Gene Details
- Gene
- St18
- Gene Name
- suppression of tumorigenicity 18
View Full Product Details