Product Details

SNP ID
rs6170712
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:54444022 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGCTGTCATGGAGTGGTCTGA[A/G]GTTGGAGTGAGCTACTATGTCAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Galnt13 PubMed Links

Gene Details

Gene
Galnt13
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173030.2 Intron NP_766618.2

View Full Product Details