Product Details
- SNP ID
-
rs13481960
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:96878987 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACTCAGAGCTCTCTTTCCTCTG[C/T]GTCTGCCCATCAACATCCTACTAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ankrd31
PubMed Links
Gene Details
- Gene
- Ankrd31
- Gene Name
- ankyrin repeat domain 31
There are no transcripts associated with this gene.
View Full Product Details