Product Details
- SNP ID
-
rs4229115
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:103937948 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAAAGGTTATATGGCCTAGCAAA[C/T]ATAGCCCCAGGTCTCTGGTCAGTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Nsf
PubMed Links
Gene Details
- Gene
- Nsf
- Gene Name
- N-ethylmaleimide sensitive fusion protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008740.4 |
|
Intron |
|
|
NP_032766.2 |
View Full Product Details