Product Details

SNP ID
rs3022933
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:143813913 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCACCCAGCAATTCCTGTTACCCCC[A/G]ACACAAGCAATCCCAGCCTGGTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Pcsk2 PubMed Links

Gene Details

Gene
Pcsk2
Gene Name
proprotein convertase subtilisin/kexin type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008792.4 2374 UTR 3 NP_032818.1

View Full Product Details