Product Details
- SNP ID
-
rs13480940
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:30608800 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACTATACTCCTGTGATGGCCCTA[A/C]CCCCATGAGTTTATGGGCAGCACAA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
Acyp2
PubMed Links
Gene Details
- Gene
- Acyp2
- Gene Name
- acylphosphatase 2, muscle type
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_029344.3 |
|
Intron |
|
|
NP_083620.1 |
View Full Product Details