Product Details

SNP ID
rs13481581
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:89400493 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGAGATGACTTTAATCTGGCT[A/G]TGCCATGGAACAGATGAGCAGCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Nrxn3 PubMed Links

Gene Details

Gene
Nrxn3
Gene Name
neurexin III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198587.3 Intron NP_001185516.2
NM_001252074.2 Intron NP_001239003.1
NM_172544.3 Intron NP_766132.2

View Full Product Details