Product Details

SNP ID
rs29348118
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:120208976 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGTCAGGGGGACCAAGTGACC[A/G]GAGGGAGACATAGTTGTCTTCTAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gm38500 PubMed Links

Gene Details

Gene
Gm38500
Gene Name
predicted gene, 38500
There are no transcripts associated with this gene.

Gene
Irak3
Gene Name
interleukin-1 receptor-associated kinase 3
There are no transcripts associated with this gene.

Gene
Tmbim4
Gene Name
transmembrane BAX inhibitor motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026617.3 Intron NP_080893.1

View Full Product Details