Product Details
- SNP ID
-
rs6249290
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:32094848 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTATGAAGGAGAGCTCTCATCC[C/T]GCGTTTCCAATCATTTGCTGATATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Iws1
PubMed Links
Gene Details
- Gene
- Iws1
- Gene Name
- IWS1 homolog (S. cerevisiae)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173441.3 |
|
Intron |
|
|
NP_775617.1 |
View Full Product Details