Product Details

SNP ID
rs4222623
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:131667917 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCACATGAAGAAAGTCCAAGA[G/C]TGGGATTATAGTGAAAATCTGTGTG
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
Ctse PubMed Links

Gene Details

Gene
Ctse
Gene Name
cathepsin E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007799.3 Intron NP_031825.2

View Full Product Details