Product Details

SNP ID
rs3667161
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:5079178 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACATGATGTACAGACCAATGAAAAA[T/C]GTTAAATTTTCAGGTTTGATTGTTT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Arid1b PubMed Links

Gene Details

Gene
Arid1b
Gene Name
AT rich interactive domain 1B (SWI-like)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085355.1 Intron NP_001078824.1

View Full Product Details