Product Details

SNP ID
rs3701665
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:35328302 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCCCTGGGCTCACAGCTTGC[A/G]CCAATGTTGGCTTGGTGATGGGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Slit3 PubMed Links

Gene Details

Gene
Slit3
Gene Name
slit homolog 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011412.3 Intron NP_035542.2

View Full Product Details