Product Details
- SNP ID
-
rs13476468
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:44618192 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATCTAGAGACATTGAACAGTCT[C/G]TTGGCTGGGGATGCAGTGTATATGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Gtdc1
PubMed Links
Gene Details
- Gene
- Gtdc1
- Gene Name
- glycosyltransferase-like domain containing 1
There are no transcripts associated with this gene.
View Full Product Details