Product Details
- SNP ID
-
rs13476283
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:184905583 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGCTGGAGCTCCCTCGGGGGAAG[C/T]GGGTCCTGTCTGGAGTGCTGGCACT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mark1
PubMed Links
Gene Details
- Gene
- Mark1
- Gene Name
- MAP/microtubule affinity regulating kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145515.2 |
1885 |
Missense Mutation |
|
|
NP_663490.2 |
View Full Product Details