Product Details
- SNP ID
-
rs13481530
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:74073741 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGACTAAACTTGGTATGGTATC[G/T]CCTCAGACAGATCTTTCTTAGCCTG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Syt16
PubMed Links
Gene Details
- Gene
- Syt16
- Gene Name
- synaptotagmin XVI
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172804.2 |
|
Intron |
|
|
NP_766392.2 |
View Full Product Details