Product Details

SNP ID
rs4225575
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:145181903 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTCTGTGCTGTCACTCAGACA[C/G]CTGTGGCATGCAGAGGCCGCCCATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Atp5j2 PubMed Links

Gene Details

Gene
Atp5j2
Gene Name
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2
There are no transcripts associated with this gene.

Gene
Cpsf4
Gene Name
cleavage and polyadenylation specific factor 4
There are no transcripts associated with this gene.

View Full Product Details