Product Details
- SNP ID
-
rs3658198
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:76567438 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCATGGGGGTTTGCATGGCTCAGG[G/A]GGGTGGGTATGTTTGAGATCTGCAA
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Abr
PubMed Links
Gene Details
- Gene
- Abr
- Gene Name
- active BCR-related gene
There are no transcripts associated with this gene.
View Full Product Details