Product Details

SNP ID
rs3090114
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:74211561 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATTGGCACCAGGTATCCAAGCTTGC[T/C]ATAGGGTGTGGTACCCCCAAAACAC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Ahrr PubMed Links

Gene Details

Gene
Ahrr
Gene Name
aryl-hydrocarbon receptor repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009644.2 4289 UTR 3 NP_033774.1
Gene
Exoc3
Gene Name
exocyst complex component 3
There are no transcripts associated with this gene.

View Full Product Details