Product Details

SNP ID
rs4230427
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:69175325 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCCTTCCTGCTGGTGGGCGC[G/A]GGGCTGGAAAGCACTGCCCACCTTA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
1700092C10Rik PubMed Links

Gene Details

Gene
1700092C10Rik
Gene Name
RIKEN cDNA 1700092C10 gene
There are no transcripts associated with this gene.

Gene
Nkx2-6
Gene Name
NK2 homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010920.2 941 UTR 3 NP_035050.2

View Full Product Details