Product Details

SNP ID
rs4229437
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:78837719 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GATAGCCTTAACGCTGAGAGTTCAT[C/G]TCTTAAACTAGTGAGAAAGGTCCAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Atp6v1d PubMed Links

Gene Details

Gene
Atp6v1d
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit D
There are no transcripts associated with this gene.

Gene
Mpp5
Gene Name
membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019579.3 2520 UTR 3 NP_062525.1

View Full Product Details