Product Details

SNP ID
rs3023432
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:10740912 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCATTGAAAAGCAAGTGGCCTGCA[A/T]CTATCCAGTCTCCAAGGGTTTAATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Clec16a PubMed Links

Gene Details

Gene
Clec16a
Gene Name
C-type lectin domain family 16, member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204229.1 Intron NP_001191158.1
NM_177562.5 Intron NP_808230.2

View Full Product Details