Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEFSEC
Entrez Gene ID
102147133
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
-
Chromosome Location
Chr.11: 88791410 - 89045726 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571771.2 XP_005571828.1 3 - 4 685 99
XM_005571774.2 XP_005571831.1 3 - 4 685 99

View Full Product Details