Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTEX1D1
Entrez Gene ID
102143718
Gene Name
Tctex1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 160575605 - 160603111 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543122.2 XP_005543179.1 4 - 5 1690 116

View Full Product Details