Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGR1
Entrez Gene ID
102128326
Gene Name
early growth response 1
Gene Aliases
-
Chromosome Location
Chr.6: 137564963 - 137568790 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557906.2 XP_005557963.2 1 - 2 597 71

View Full Product Details