Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXD8
Entrez Gene ID
102146552
Gene Name
homeobox D8
Gene Aliases
-
Chromosome Location
Chr.12: 65095941 - 65098011 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574929.2 XP_005574986.1 1 - 2 288 71

View Full Product Details