Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTR1
Entrez Gene ID
102141078
Gene Name
endosome associated trafficking regulator 1
Gene Aliases
SDCCAG3
Chromosome Location
Chr.15: 1938570 - 1950727 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580418.1 XP_005580475.1 7 - 8 1191 66
XM_005580419.2 XP_005580476.1 7 - 8 1150 66
XM_005580420.2 XP_005580477.1 6 - 7 1121 66
XM_005580421.2 XP_005580478.1 5 - 6 971 66

View Full Product Details