Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVX1
Entrez Gene ID
102131087
Gene Name
even-skipped homeobox 1
Gene Aliases
-
Chromosome Location
Chr.3: 93479927 - 93485031 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549920.2 XP_005549977.1 1 - 2 756 96

View Full Product Details