Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOE
Entrez Gene ID
102138471
Gene Name
apolipoprotein E
Gene Aliases
Apo-E
Chromosome Location
Chr.19: 45651596 - 45655521 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589554.2 XP_005589611.2 3 - 4 316 58

View Full Product Details