Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAT1
Entrez Gene ID
102146166
Gene Name
spermidine/spermine N1-acetyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.X: 21738200 - 21741351 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593176.2 XP_005593233.1 2 - 3 483 92
XM_005593177.2 XP_005593234.1 3 - 4 481 92

View Full Product Details