Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GEM
Entrez Gene ID
102131323
Gene Name
GTP binding protein overexpressed in skeletal muscle
Gene Aliases
-
Chromosome Location
Chr.8: 95383552 - 95396870 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563707.2 XP_005563764.1 3 - 4 551 111

View Full Product Details