Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL46
Entrez Gene ID
102124639
Gene Name
mitochondrial ribosomal protein L46
Gene Aliases
-
Chromosome Location
Chr.7: 68663057 - 68669884 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560428.2 XP_005560485.2 2 - 3 472 99

View Full Product Details