Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF10
Entrez Gene ID
102125677
Gene Name
Kruppel like factor 10
Gene Aliases
-
Chromosome Location
Chr.8: 103804089 - 103810981 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563841.2 XP_005563898.1 3 - 4 1477 105

View Full Product Details