Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRKB
Entrez Gene ID
102118351
Gene Name
TP53RK binding protein
Gene Aliases
-
Chromosome Location
Chr.13: 36185235 - 36193290 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575585.2 XP_005575642.1 2 - 3 575 111
XM_005575586.2 XP_005575643.1 2 - 3 587 111

View Full Product Details