Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAH
Entrez Gene ID
102125144
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
-
Chromosome Location
Chr.7: 58620240 - 58656021 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560246.1 XP_005560303.1 3 - 4 292 59
XM_005560247.2 XP_005560304.1 3 - 4 280 59
XM_005560248.1 XP_005560305.1 2 - 3 175 59
XM_005560249.2 XP_005560306.1 2 - 3 170 59

View Full Product Details