Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FIGN
Entrez Gene ID
102140122
Gene Name
fidgetin, microtubule severing factor
Gene Aliases
-
Chromosome Location
Chr.12: 52432845 - 52563243 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573329.2 XP_005573386.1 1 - 2 347 80

View Full Product Details