Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOS
Entrez Gene ID
102119607
Gene Name
otospiralin
Gene Aliases
-
Chromosome Location
Chr.12: 130018251 - 130020263 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574810.2 XP_005574867.1 3 - 4 617 147
XM_005574812.2 XP_005574869.1 3 - 4 537 147

View Full Product Details