Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK11
Entrez Gene ID
102143615
Gene Name
serine/threonine kinase 11
Gene Aliases
-
Chromosome Location
Chr.19: 980968 - 1003999 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005587363.2 XP_005587420.1 3 - 4 914 104
XM_005587364.2 XP_005587421.1 3 - 4 815 104
XM_015440751.1 XP_015296237.1 3 - 4 815 104
GenBank mRNA AB220526.1 - 1 - 2 101 104

View Full Product Details