Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
102139170
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
Chromosome Location
Chr.15: 44507648 - 44514639 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437061.1 XP_015292547.1 6 - 7 1162 92

View Full Product Details