Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENTPD2
Entrez Gene ID
102119518
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
-
Chromosome Location
Chr.15: 1241228 - 1246908 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580352.2 XP_005580409.1 3 - 4 461 112

View Full Product Details