Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRNA6
Entrez Gene ID
102122218
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
-
Chromosome Location
Chr.8: 43781066 - 43796988 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563230.1 XP_005563287.1 4 - 5 819 104

View Full Product Details