Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHOD
Entrez Gene ID
102144069
Gene Name
ras homolog family member D
Gene Aliases
-
Chromosome Location
Chr.14: 7421890 - 7438600 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577105.2 XP_005577162.1 3 - 4 409 89
XM_015434387.1 XP_015289873.1 4 - 5 420 89

View Full Product Details