Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE2
Entrez Gene ID
102115013
Gene Name
jade family PHD finger 2
Gene Aliases
PHF15
Chromosome Location
Chr.6: 133578940 - 133637170 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557789.2 XP_005557846.2 4 - 5 564 134
XM_005557790.2 XP_005557847.1 4 - 5 565 134
XM_005557791.2 XP_005557848.1 4 - 5 566 134
XM_005557792.2 XP_005557849.1 4 - 5 566 134
XM_005557795.2 XP_005557852.1 4 - 5 669 134
XM_005557796.2 XP_005557853.1 4 - 5 430 134
XM_005557798.2 XP_005557855.1 4 - 5 431 134
XM_005557799.2 XP_005557856.1 4 - 5 440 134
XM_015451895.1 XP_015307381.1 5 - 6 669 134
XM_015451896.1 XP_015307382.1 4 - 5 669 134
XM_015451897.1 XP_015307383.1 3 - 4 1843 134

View Full Product Details