Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
102140769
Gene Name
matrix remodeling associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 962083 - 996431 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592897.2 XP_005592954.1 5 - 6 6319 104

View Full Product Details