Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
102127785
Gene Name
syntrophin gamma 2
Gene Aliases
-
Chromosome Location
Chr.13: 109848173 - 110224038 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576652.2 XP_005576709.1 12 - 13 1170 99

View Full Product Details