Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPEP2
Entrez Gene ID
102128559
Gene Name
dipeptidase 2
Gene Aliases
-
Chromosome Location
Chr.20: 56192136 - 56200239 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442779.1 XP_015298265.1 9 - 10 1590 130

View Full Product Details