Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP1
Entrez Gene ID
102133951
Gene Name
receptor accessory protein 1
Gene Aliases
-
Chromosome Location
Chr.13: 23142948 - 23210901 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575375.2 XP_005575432.1 5 - 6 491 111

View Full Product Details