Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN33
Entrez Gene ID
102121102
Gene Name
tetraspanin 33
Gene Aliases
-
Chromosome Location
Chr.3: 161794125 - 161820543 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550736.2 XP_005550793.1 1 - 2 176 75
XM_005550737.2 XP_005550794.1 1 - 2 161 75
XM_015448024.1 XP_015303510.1 1 - 2 176 75

View Full Product Details