Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXF2
Entrez Gene ID
102135675
Gene Name
forkhead box F2
Gene Aliases
-
Chromosome Location
Chr.4: 169666562 - 169672079 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554232.2 XP_005554289.2 1 - 2 1065 80

View Full Product Details