Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFKBIL1
Entrez Gene ID
102136863
Gene Name
NFKB inhibitor like 1
Gene Aliases
-
Chromosome Location
Chr.4: 139323268 - 139335838 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553552.2 XP_005553609.1 3 - 4 681 66
XM_005553555.2 XP_005553612.1 4 - 5 916 66
XM_005553556.2 XP_005553613.1 4 - 5 620 66
XM_005553557.2 XP_005553614.1 3 - 4 809 66

View Full Product Details